siRNA Details
| Gene Symbol: Med13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080931.1 | NP_001074400.1 | 6 | 992 | ||
| GenBank mRNA | AK083472.1 | - | 3 | 2617 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med13| Gene Symbol: Med13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080931.1 | NP_001074400.1 | 6 | 992 | ||
| GenBank mRNA | AK083472.1 | - | 3 | 2617 | ||