siRNA Details
| Gene Symbol: D130037M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083877.1 | - | 244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D130037M23Rik| Gene Symbol: D130037M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083877.1 | - | 244 | |||