siRNA Details
| Gene Symbol: Mcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085373.1 | NP_001078842.1 | 11 | 1743 | ||
| NM_001085374.1 | NP_001078843.1 | 9 | 1711 | |||
| GenBank mRNA | AK054528.1 | - | 9 | 1713 | ||
| AK158266.1 | - | 11 | 1744 | |||

Mcc
Genomic Map