siRNA Details
| Gene Symbol: Arhgef37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177828.4 | NP_808496.2 | 12 | 1873 | ||
| GenBank mRNA | AB199795.1 | - | 11 | 1723 | ||
| AK030242.1 | - | 8 | 1700 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef37| Gene Symbol: Arhgef37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177828.4 | NP_808496.2 | 12 | 1873 | ||
| GenBank mRNA | AB199795.1 | - | 11 | 1723 | ||
| AK030242.1 | - | 8 | 1700 | |||