siRNA Details
| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081307.1 | NP_001074776.1 | 13, 14 | 2001 | ||
| GenBank mRNA | AK141375.1 | - | 15, 16 | 2216 | ||
| BC145746.1 | - | 13, 14 | 1912 | 40131049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r12b| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081307.1 | NP_001074776.1 | 13, 14 | 2001 | ||
| GenBank mRNA | AK141375.1 | - | 15, 16 | 2216 | ||
| BC145746.1 | - | 13, 14 | 1912 | 40131049 | ||