siRNA Details
| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081307.1 | NP_001074776.1 | 14 | 2034 | ||
| GenBank mRNA | AK039445.1 | - | 2 | 95 | ||
| AK141375.1 | - | 16 | 2249 | |||
| BC145746.1 | - | 14 | 1945 | 40131049 | ||

Ppp1r12b
Genomic Map