siRNA Details
Gene Symbol: Ppp1r12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081307.1 | NP_001074776.1 | 10 | 1440 | ||
GenBank mRNA | AK046012.1 | - | 10 | 1429 | ||
AK046167.1 | - | 10 | 1429 | |||
AK080855.1 | - | 10 | 1429 | |||
AK141375.1 | - | 10 | 1429 | |||
BC145746.1 | - | 10 | 1351 | 40131049 |
