siRNA Details
| Gene Symbol: Myo3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177376.4 | NP_796350.2 | 35 | 3916 | ||
| GenBank mRNA | AK040119.1 | - | 10 | 1069 | ||
| AY830392.2 | - | 34 | 3887 | |||
| AY830393.2 | - | 33 | 3755 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo3b| Gene Symbol: Myo3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177376.4 | NP_796350.2 | 35 | 3916 | ||
| GenBank mRNA | AK040119.1 | - | 10 | 1069 | ||
| AY830392.2 | - | 34 | 3887 | |||
| AY830393.2 | - | 33 | 3755 | |||