siRNA Details
| Gene Symbol: Accs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290782.1 | NP_001277711.1 | 5 | 624 | ||
| NM_183220.3 | NP_899043.1 | 5 | 679 | |||
| GenBank mRNA | AK011867.1 | - | 5 | 528 | ||
| AK040509.1 | - | 5 | 521 | |||
| BC039569.1 | - | 5 | 656 | 1449080 | ||

Accs
Genomic Map