siRNA Details
| Gene Symbol: Zfp335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199027.2 | NP_950192.2 | 5 | 1015 | ||
| GenBank mRNA | AK157577.1 | - | 5 | 843 | ||
| BC062921.1 | - | 5 | 1015 | |||
| BC151013.1 | - | 4 | 752 | 9087926 | ||
| BC157954.1 | - | 4 | 752 | 9088127 | ||
| BC172135.1 | - | 4 | 752 | |||

Zfp335
Genomic Map