siRNA Details
| Gene Symbol: Aknad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177859.3 | NP_808527.2 | 2 | 449 | ||
| GenBank mRNA | AK029552.1 | - | 2 | 327 | ||
| AK164420.1 | - | 2 | 450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aknad1| Gene Symbol: Aknad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177859.3 | NP_808527.2 | 2 | 449 | ||
| GenBank mRNA | AK029552.1 | - | 2 | 327 | ||
| AK164420.1 | - | 2 | 450 | |||