siRNA Details
| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177861.4 | NP_808529.2 | 13 | 1383 | ||
| GenBank mRNA | AK045429.1 | - | 13 | 1384 | ||
| AK077246.1 | - | 13 | 1430 | |||
| AK133059.1 | - | 13 | 1360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem67| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177861.4 | NP_808529.2 | 13 | 1383 | ||
| GenBank mRNA | AK045429.1 | - | 13 | 1384 | ||
| AK077246.1 | - | 13 | 1430 | |||
| AK133059.1 | - | 13 | 1360 | |||