siRNA Details
| Gene Symbol: Catsper4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130030.1 | NP_001123502.1 | 6 | 645 | ||
| NM_177866.4 | NP_808534.1 | 8 | 864 | |||
| GenBank mRNA | AK077145.1 | - | 8 | 851 | ||
| BC108974.2 | - | 8 | 844 | 40047424 | ||
| BC108975.1 | - | 8 | 844 | 40047425 | ||
| EU781510.1 | - | 7 | 741 | |||
| EU781511.1 | - | 6 | 620 | |||

Catsper4
Genomic Map