siRNA Details
| Gene Symbol: Gm438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126316.1 | NP_001119788.1 | 1, 2 | 235 | ||
| GenBank mRNA | BB630382.1 | - | 1, 2 | 235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm438| Gene Symbol: Gm438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126316.1 | NP_001119788.1 | 1, 2 | 235 | ||
| GenBank mRNA | BB630382.1 | - | 1, 2 | 235 | ||