siRNA Details
| Gene Symbol: Vps37b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177876.4 | NP_808544.1 | 3 | 399 | ||
| GenBank mRNA | AK082658.1 | - | 3 | 360 | ||
| AK149893.1 | - | 3 | 366 | |||
| AK172114.1 | - | 3 | 360 | |||
| AK172337.1 | - | 3 | 360 | |||
| BC026744.1 | - | 3 | 400 | 4163734 | ||

Vps37b
Genomic Map