siRNA Details
| Gene Symbol: Gal3st4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033416.2 | NP_001028588.1 | 3 | 1921 | ||
| GenBank mRNA | AK132558.1 | - | 3 | 1922 | ||
| AK138993.1 | - | 4 | 586 | |||
| AK162468.1 | - | 1 | 146 | |||
| Gene Symbol: 6330418K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031849.1 | - | 2 | 772 | ||

Gal3st4
Genomic Map