siRNA Details
| Gene Symbol: Pon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183308.2 | NP_899131.1 | 1, 2 | 161 | ||
| GenBank mRNA | AK007350.1 | - | 1, 2 | 129 | ||
| AK146311.1 | - | 1, 2 | 129 | |||
| AK152351.1 | - | 1, 2 | 112 | |||
| AK171926.1 | - | 1, 2 | 119 | |||
| BC021887.1 | - | 1, 2 | 118 | 3154583 | ||
| BC037140.1 | - | 1, 2 | 103 | 5027719 | ||
| BC055896.1 | - | 1, 2 | 161 | 3482584 | ||
| BC062200.1 | - | 1, 2 | 152 | 6399484 | ||

Pon2
Genomic Map