siRNA Details
Gene Symbol: Fam188b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142781.1 | NP_001136253.1 | 8 | 1404 | ||
NM_177883.4 | NP_808551.2 | 8 | 1404 | |||
GenBank mRNA | AK082855.1 | - | 8 | 1405 | ||
AK142385.1 | - | 8 | 1322 | |||
AK161304.1 | - | 8 | 1283 | |||
BC118939.1 | - | 8 | 1398 | |||
BC118940.1 | - | 8 | 1398 | 40092089 |
