siRNA Details
| Gene Symbol: Gm766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145390.1 | NP_001138862.1 | 2, 3 | 412 | ||
| GenBank mRNA | AK144423.1 | - | 449 | |||
| BC151094.1 | - | 2, 3 | 412 | 9088007 | ||
| BC151096.1 | - | 2, 3 | 412 | 9088009 | ||
| BC172155.1 | - | 2, 3 | 412 | 9054825 | ||

Gm766
Genomic Map