siRNA Details
| Gene Symbol: Far2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178797.3 | NP_848912.1 | 12 | 1609 | ||
| GenBank mRNA | AK038584.1 | - | 12 | 1562 | ||
| AK043388.1 | - | 12 | 1609 | |||
| AK043589.1 | - | 12 | 1609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Far2| Gene Symbol: Far2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178797.3 | NP_848912.1 | 12 | 1609 | ||
| GenBank mRNA | AK038584.1 | - | 12 | 1562 | ||
| AK043388.1 | - | 12 | 1609 | |||
| AK043589.1 | - | 12 | 1609 | |||