siRNA Details
Gene Symbol: Tmem150b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142792.1 | NP_001136264.1 | 1, 2 | 163 | ||
NM_177887.4 | NP_808555.1 | 1, 2 | 332 | |||
GenBank mRNA | AK041846.1 | - | 1, 2 | 333 | ||
BC022651.1 | - | 1, 2 | 163 | 4216812 |
