siRNA Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198631.2 | NP_941033.2 | 3 | 369 | ||
| GenBank mRNA | AK138078.1 | - | 2 | 2187 | ||
| AK151148.1 | - | 4 | 421 | |||
| AK151306.1 | - | 4 | 421 | |||
| BC057627.1 | - | 3 | 314 | 6412890 | ||
| BY167917.1 | - | 4 | 418 | |||
| CJ101946.1 | - | 3 | 374 | |||

Zc3h4
Genomic Map