siRNA Details
| Gene Symbol: Tmem145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183311.2 | NP_899134.2 | 7, 8 | 666 | ||
| GenBank mRNA | AK081054.1 | - | 7, 8 | 667 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem145| Gene Symbol: Tmem145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183311.2 | NP_899134.2 | 7, 8 | 666 | ||
| GenBank mRNA | AK081054.1 | - | 7, 8 | 667 | ||