siRNA Details
| Gene Symbol: Hcn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081192.1 | NP_001074661.1 | 8 | 3441 | ||
| GenBank mRNA | BC158106.1 | - | 8 | 3541 | 9088182 | |
| BC158108.1 | - | 8 | 3541 | 9088183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcn4| Gene Symbol: Hcn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081192.1 | NP_001074661.1 | 8 | 3441 | ||
| GenBank mRNA | BC158106.1 | - | 8 | 3541 | 9088182 | |
| BC158108.1 | - | 8 | 3541 | 9088183 | ||