siRNA Details
| Gene Symbol: Slc9a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177909.5 | NP_808577.3 | 1 | 239 | ||
| GenBank mRNA | AK037058.1 | - | 1 | 223 | ||
| AK156494.1 | - | 1 | 200 | |||
| AK172687.1 | - | 1 | 211 | |||
| BC099942.1 | - | 1 | 213 | 30103308 | ||
| BC153825.1 | - | 1 | 217 | 40093280 | ||

Slc9a9
Genomic Map