siRNA Details
| Gene Symbol: Dgkk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177914.3 | NP_808582.3 | 3040 | |||
| GenBank mRNA | BC075627.1 | - | 3040 | 6852516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgkk| Gene Symbol: Dgkk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177914.3 | NP_808582.3 | 3040 | |||
| GenBank mRNA | BC075627.1 | - | 3040 | 6852516 | ||