siRNA Details
| Gene Symbol: Card9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037747.1 | NP_001032836.1 | 5 | 903 | ||
| GenBank mRNA | BC151023.1 | - | 5 | 1111 | 9087936 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Card9| Gene Symbol: Card9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037747.1 | NP_001032836.1 | 5 | 903 | ||
| GenBank mRNA | BC151023.1 | - | 5 | 1111 | 9087936 | |