siRNA Details
| Gene Symbol: Proser3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183321.1 | NP_899144.1 | 10 | 1235 | ||
| GenBank mRNA | BC053749.1 | - | 10 | 1235 | 6331972 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Proser3| Gene Symbol: Proser3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183321.1 | NP_899144.1 | 10 | 1235 | ||
| GenBank mRNA | BC053749.1 | - | 10 | 1235 | 6331972 | |