siRNA Details
| Gene Symbol: Trim75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033429.2 | NP_001028601.1 | 2 | 543 | ||
| GenBank mRNA | AK136018.1 | - | 2 | 545 | ||
| BC139219.1 | - | 1 | 250 | 8862241 | ||
| BC139221.1 | - | 1 | 250 | 8862243 | ||

Trim75
Genomic Map