siRNA Details
| Gene Symbol: Ttc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290563.1 | NP_001277492.1 | 4, 5 | 611 | ||
| NM_001290564.1 | NP_001277493.1 | 4, 5 | 666 | |||
| NM_001290566.1 | NP_001277495.1 | 3, 4 | 364 | |||
| NM_001290567.1 | NP_001277496.1 | 4, 5 | 412 | |||
| NM_177384.3 | NP_796358.2 | 4, 5 | 611 | |||
| GenBank mRNA | AK028040.1 | - | 4, 5 | 666 | ||
| AK089384.1 | - | 3, 4 | 644 | |||
| AK133273.1 | - | 4, 5 | 389 | |||
| AK160435.1 | - | 4, 5 | 362 | |||
| AK164721.1 | - | 4, 5 | 565 | |||
| BC140991.1 | - | 3, 4 | 355 | 9055603 | ||
| BC145064.1 | - | 4, 5 | 403 | 9053612 | ||

Ttc16
Genomic Map