siRNA Details
| Gene Symbol: Ttc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290564.1 | NP_001277493.1 | 15 | 2617 | ||
| NM_001290566.1 | NP_001277495.1 | 14 | 2315 | |||
| NM_001290567.1 | NP_001277496.1 | 15 | 2363 | |||
| NM_177384.3 | NP_796358.2 | 15 | 2562 | |||
| GenBank mRNA | AK028040.1 | - | 15 | 2617 | ||
| AK133273.1 | - | 15 | 2340 | |||
| BC140991.1 | - | 14 | 2306 | 9055603 | ||
| BC145064.1 | - | 15 | 2354 | 9053612 | ||

Ttc16
Genomic Map