siRNA Details
| Gene Symbol: Cntln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175275.4 | NP_780484.2 | 3 | 623 | ||
| NM_177385.4 | NP_796359.1 | 3 | 623 | |||
| GenBank mRNA | AK031604.1 | - | 3 | 529 | ||
| AK052558.1 | - | 3 | 543 | |||
| AK165111.1 | - | 2 | 96 | |||
| BC125622.1 | - | 3 | 498 | |||
| BC125624.1 | - | 3 | 498 | |||

Cntln
Genomic Map