siRNA Details
| Gene Symbol: Akap17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081956.1 | NP_001075425.1 | 7 | 2512 | ||
| GenBank mRNA | AK048769.1 | - | 1 | 508 | ||
| BC120735.1 | - | 1 | 508 | |||
| BC131917.1 | - | 1 | 508 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap17b| Gene Symbol: Akap17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081956.1 | NP_001075425.1 | 7 | 2512 | ||
| GenBank mRNA | AK048769.1 | - | 1 | 508 | ||
| BC120735.1 | - | 1 | 508 | |||
| BC131917.1 | - | 1 | 508 | |||