siRNA Details
| Gene Symbol: Zfp780b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081021.1 | NP_001074490.1 | 6 | 2171 | ||
| GenBank mRNA | AK045524.1 | - | 6 | 2162 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp780b| Gene Symbol: Zfp780b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081021.1 | NP_001074490.1 | 6 | 2171 | ||
| GenBank mRNA | AK045524.1 | - | 6 | 2162 | ||