siRNA Details
| Gene Symbol: Tas2r117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207021.1 | NP_996904.1 | 1 | 481 | ||
| GenBank mRNA | BC132206.1 | - | 1 | 564 | 40130483 | |
| BC132208.1 | - | 1 | 564 | 40130485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r117| Gene Symbol: Tas2r117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207021.1 | NP_996904.1 | 1 | 481 | ||
| GenBank mRNA | BC132206.1 | - | 1 | 564 | 40130483 | |
| BC132208.1 | - | 1 | 564 | 40130485 | ||