siRNA Details
| Gene Symbol: Adam32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293693.1 | NP_001280622.1 | 6 | 461 | ||
| NM_153397.2 | NP_700446.2 | 6 | 461 | |||
| GenBank mRNA | AF513715.1 | - | 6 | 444 | ||
| AK005759.1 | - | 6 | 452 | |||
| BC060983.1 | - | 6 | 458 | 6772485 | ||

Adam32
Genomic Map