siRNA Details
| Gene Symbol: Pcdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198117.2 | NP_932785.1 | 1 | 1104 | ||
| GenBank mRNA | AY013760.1 | - | 1 | 991 | ||
| BC030897.1 | - | 1 | 1104 | |||
| BC141390.1 | - | 1 | 1085 | 9056003 | ||
| EU854475.1 | - | 1 | 94 | |||

Pcdha2
Genomic Map