siRNA Details
| Gene Symbol: Ltv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181470.4 | NP_852135.1 | 5 | 531 | ||
| GenBank mRNA | AK011491.1 | - | 5 | 517 | ||
| BC067062.1 | - | 5 | 531 | 30538957 | ||
| BC069962.1 | - | 5 | 525 | 5038459 | ||
| U01139.1 | - | 2 | 106 | |||

Ltv1
Genomic Map