siRNA Details
| Gene Symbol: Opn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181753.4 | NP_861418.2 | 3 | 401 | ||
| GenBank mRNA | AY288426.1 | - | 3 | 388 | ||
| AY318865.1 | - | 3 | 402 | |||
| BC125611.1 | - | 3 | 483 | 40130126 | ||
| BC132010.1 | - | 3 | 483 | 40130287 | ||
More Information
Other Species Targeted
Set Membership: |
|

Opn5
Genomic Map