siRNA Details
| Gene Symbol: Opn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181753.4 | NP_861418.2 | 6 | 1020 | ||
| GenBank mRNA | AK039525.1 | - | 2 | 211 | ||
| AY288426.1 | - | 6 | 1007 | |||
| AY318865.1 | - | 6 | 1021 | |||
| BC125611.1 | - | 6 | 1102 | 40130126 | ||
| BC132010.1 | - | 6 | 1102 | 40130287 | ||
More Information
Set Membership: |
|

Opn5
Genomic Map