siRNA Details
| Gene Symbol: Opn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181753.4 | NP_861418.2 | 4 | 682 | ||
| GenBank mRNA | AY288426.1 | - | 4 | 669 | ||
| AY318865.1 | - | 4 | 683 | |||
| BC125611.1 | - | 4 | 764 | 40130126 | ||
| BC132010.1 | - | 4 | 764 | 40130287 | ||
More Information
Set Membership: |
|

Opn5
Genomic Map