siRNA Details
Gene Symbol: Gpr141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181754.4 | NP_861419.3 | 5 | 1174 | ||
GenBank mRNA | AK154688.1 | - | 5 | 1175 | ||
AY255553.1 | - | 1 | 144 | |||
AY288427.1 | - | 1 | 663 | |||
BC125585.1 | - | 1 | 747 | 40130100 | ||
BC125587.1 | - | 1 | 747 | 40130102 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
