siRNA Details
| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181990.2 | NP_871790.2 | 1 | 589 | ||
| GenBank mRNA | AY241873.1 | - | 366 | |||
| AY241874.1 | - | 800 | ||||
| BC145587.1 | - | 1 | 589 | 9054153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtm1| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181990.2 | NP_871790.2 | 1 | 589 | ||
| GenBank mRNA | AY241873.1 | - | 366 | |||
| AY241874.1 | - | 800 | ||||
| BC145587.1 | - | 1 | 589 | 9054153 | ||