siRNA Details
| Gene Symbol: Cklf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037841.3 | NP_001032930.1 | 3 | 1578 | ||
| GenBank mRNA | AK032759.1 | - | 3 | 1508 | ||
| AK086063.1 | - | 3 | 1511 | |||
| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181990.2 | NP_871790.2 | 1 | 235 | ||
| GenBank mRNA | AY241873.1 | - | 144 | |||
| AY241874.1 | - | 446 | ||||
| BC145587.1 | - | 1 | 235 | 9054153 | ||

Cklf
Genomic Map