siRNA Details
| Gene Symbol: Myo18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291212.1 | NP_001278141.1 | 1 | 664 | ||
| NM_001291213.1 | NP_001278142.1 | 1 | 664 | |||
| NM_011586.3 | NP_035716.1 | 1 | 664 | |||
| GenBank mRNA | AB026497.1 | - | 1 | 634 | ||
| AK146852.1 | - | 2 | 744 | |||
| BC138366.1 | - | 1 | 606 | 8861386 | ||
| BC145257.1 | - | 1 | 606 | |||

Myo18a
Genomic Map