siRNA Details
| Gene Symbol: Zranb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207302.1 | NP_997185.1 | 8, 9 | 1903 | ||
| GenBank mRNA | BC145520.1 | - | 5, 6 | 1516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zranb1| Gene Symbol: Zranb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207302.1 | NP_997185.1 | 8, 9 | 1903 | ||
| GenBank mRNA | BC145520.1 | - | 5, 6 | 1516 | ||