siRNA Details
| Gene Symbol: Fastkd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146084.1 | NP_001139556.1 | 3 | 2156 | ||
| NM_198176.2 | NP_937819.2 | 3 | 2193 | |||
| GenBank mRNA | AK143929.1 | - | 3 | 2132 | ||
| AK220420.1 | - | 1 | 1757 | |||
| BC048956.1 | - | 1 | 1195 | |||
| BC052898.1 | - | 3 | 2113 | 30059761 | ||

Fastkd5
Genomic Map