siRNA Details
| Gene Symbol: Fastkd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146084.1 | NP_001139556.1 | 3 | 1496 | ||
| NM_198176.2 | NP_937819.2 | 3 | 1533 | |||
| GenBank mRNA | AK143929.1 | - | 3 | 1472 | ||
| AK220420.1 | - | 1 | 1097 | |||
| BC048956.1 | - | 1 | 535 | |||
| BC052898.1 | - | 3 | 1453 | 30059761 | ||
| Gene Symbol: Ubox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK170024.1 | - | 2 | 1391 | ||

Fastkd5
Genomic Map