siRNA Details
| Gene Symbol: Lin28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031772.2 | NP_001026942.1 | 5 | 902 | ||
| GenBank mRNA | BC085193.1 | - | 3 | 645 | ||
| BC089037.1 | - | 5 | 893 | 30690977 | ||
| DQ127225.1 | - | 5 | 902 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lin28b| Gene Symbol: Lin28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031772.2 | NP_001026942.1 | 5 | 902 | ||
| GenBank mRNA | BC085193.1 | - | 3 | 645 | ||
| BC089037.1 | - | 5 | 893 | 30690977 | ||
| DQ127225.1 | - | 5 | 902 | |||