siRNA Details
| Gene Symbol: Sec14l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029937.2 | NP_001025108.1 | 8 | 694 | ||
| GenBank mRNA | AK136060.1 | - | 2 | 47 | ||
| BC138982.1 | - | 8 | 694 | 8862004 | ||
| BC138985.1 | - | 8 | 694 | 8862007 | ||
| BC145485.1 | - | 8 | 737 | |||

Sec14l3
Genomic Map